Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Nitrile Gloves

Manufacturer:  Honeywell™ LA111EB/8

 View more versions of this product

Catalog No. 11-393-63B



Add to cart

Description

Description

  • 100% nitrile content gives superior resistance to cuts, snags, abrasions and punctures
  • Available in blue or green with smooth or flocked interior
  • 15 mil or 11 mil thickness
  • Curved-hand shape with straight cuff edge
  • For handling fats, petroleum products, and many chemicals
  • USDA accepted for food processing

Order Info

Available in packs of 12 pairs. Length, 13 in. (33cm).

Specifications

Specifications

Safety
Straight
Nitrile
8
Blue
33 cm
Sand patch
aerospace and automotive degreasing, automotive assembling and painting, chemical handling, food processing, graphic arts and print shop cleanup
Curved-hand shape
11 mil
13 in.
21 CFR, USDA, FDA
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit