Learn More
Honeywell™ North™ Nitrile Gloves
$16.90 - $604.00
Specifications
Certifications/Compliance 21 CFR, USDA, FDA
Coating Material Sand patch
Length (English) 13 in.
Length (Metric) 33 cm
Material Nitrile
View More Specs
Products 22
Catalog Number Mfr. No. Color Size Palm Type Thickness (English) Price Quantity & Availability  
Catalog Number Mfr. No. Color Size Palm Type Thickness (English) Price Quantity & Availability  
19-025-708
View Documents
Honeywell™
LA102G/11
Green 11 Curved-hand shape 11 mil
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025707
View Documents
Honeywell™
LA102G/10XL
Green 10 Curved-hand shape 11 mil
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025-704
View Documents
Honeywell™
LA102G/7S
Green 7 Curved-hand shape 11 mil
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025-705
View Documents
Honeywell™
LA102G/8M
Green 8 Curved-hand shape 11 mil
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025-706
View Documents
Honeywell™
LA102G/9L
Green 9 Curved-hand shape 11 mil
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
11-393-63D
View Documents
Honeywell™
LA111EB/10
Blue 10 Curved-Hand Shape 11 mil
Dozen of 12 for $38.00
Case of 12 DZ for $456.00
Add to cart
11-393-63B
View Documents
Honeywell™
LA111EB/8
Blue 8 Curved-hand shape 11 mil
Dozen of 12 for $33.00
Case of 12 DZ for $396.00
Add to cart
11-393-64E
View Documents
Honeywell™
LA111EBFL/11
Blue 11 Curved-hand shape 15mil
Dozen of 12 for $48.50
Case of 12 DZ for $582.00
Add to cart
11-393-64D
View Documents
Honeywell™
LA111EBFL/10
Blue 10 Curved-hand shape 15mil
Dozen of 12 for $18.59
Case of 12 DZ for $618.00
Add to cart
11-393-64B
View Documents
Honeywell™
LA111EBFL/8
Blue 8 Curved-hand shape 15mil
Dozen of 12 for $16.90
Case of 12 DZ for $561.00
Add to cart
19-025-723
View Documents
Honeywell™
LA132G/11XXL
Green 11 Curved-hand shape 15mil
Case of 144 for $541.00
Add to cart
19-025719
View Documents
Honeywell™
LA132G/7S
Green 7 Curved-hand shape 15mil
Case of 144 for $541.00
Add to cart
11-393-64C
View Documents
Honeywell™
LA111EBFL/9
Blue 9 Curved-hand shape 15mil
Dozen of 12 for $46.75
Case of 12 DZ for $561.00
Add to cart
19-025-720
View Documents
Honeywell™
LA132G/8M
Green 8 Curved-hand shape 15mil
Case of 144 for $541.00
Add to cart
19-025721
View Documents
Honeywell™
LA132G/9L
Green 9 Curved-hand shape 15mil
Case of 144 for $541.00
Add to cart
19-025722
View Documents
Honeywell™
LA132G/10XL
Green 10 Curved-hand shape 15mil
Case of 144 for $541.00
Add to cart
17-987-506
View Documents
Honeywell™
LA172G/9L
Green 9 Resistance to cuts, snags and abrasions 11 mil
Case of 144 for $604.00
Add to cart
17-987-507
View Documents
Honeywell™
LA172G/10XL
Green 10 Resistance to cuts, snags and abrasions 11 mil
Case of 144 for $604.00
Add to cart
11-393-63E
View Documents
Honeywell™
LA111EB/11
Blue 11 Curved-Hand Shape 11 mil This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
11-393-63C
View Documents
Honeywell™
LA111EB/9
Blue 9 Curved-hand shape 11 mil This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
11-393-63A
View Documents
Honeywell™
LA111EB/7
Blue 7 Curved-hand shape 11 mil This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
11-393-64A
View Documents
Honeywell™
LA111EBFL/7
Blue 7 Curved-hand shape 15mil This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • 100% nitrile content gives superior resistance to cuts, snags, abrasions and punctures
  • Available in blue or green with smooth or flocked interior
  • 15 mil or 11 mil thickness
  • Curved-hand shape with straight cuff edge
  • For handling fats, petroleum products, and many chemicals
  • USDA accepted for food processing
Order Info
Available in packs of 12 pairs. Length, 13 in. (33cm).
Specifications
Specifications
21 CFR, USDA, FDA
13 in.
Nitrile
Straight
Sand patch
33 cm
aerospace and automotive degreasing, automotive assembling and painting, chemical handling, food processing, graphic arts and print shop cleanup