North Non-Aspirin Pain Relievers

Provide quick relief from headaches, minor pain and discomfort

Manufacturer: Honeywell Safety Products 161987

 View more versions of this product

Catalog No. 17-986-428

  Check Availability
Add to Cart

    Specifications

    Active Ingredients Acetaminophen 500mg
    Inactive Ingredients Povidone, Pregelatinized Starch, Stearic Acid
    For Use With (Application) Provide quick relief from headaches, minor pain and discomfort
    Format Tablet
    Quantity 250 Pack
    Storage Requirements 15 to 30°C
    Product Type Non-Aspirin Pain Reliever Aypanal Tablet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit