North Nonadherent Bandages
Won't stick to wounds
Manufacturer: Honeywell Safety Products 063634
 View more versions of this product
Catalog No. 19-310-033
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Size 7.62 x 10.16cm (3 x 4 in.)
    For Use With (Application) For use with industrial first-aid stations for extra protection from minor cuts and abrasions
    Product Type Non-adherent Pad
    Quantity 100 Box
    Sterile Yes
  • Description & Specifications
Related Products
Product Content Feedback