Learn More
Honeywell North™ NorthFlex Light Task Plus II™ Gloves
PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life
Manufacturer:  Honeywell NFD15/7S
 View more versions of this product
Catalog No. 19-150-1002
  Check Availability
Add to cart
Specifications
Description
North NorthFlex Light Task II Polyurethane Coated Dyneema Gloves
  • PU (polyurethane) palm coating is light for comfort and maximum tactility, but provides high abrasion and puncture resistance
  • Glove conforms to the shape of the hand and provides minimal resistance to hand movement, thus greatly reducing fatigue
  • The back of the glove is un-coated, for ventilation and wearer comfort
  • Contains no reaction causing, natural rubber proteins
  • Cuff over-stitching is color-coded by size to aid sorting after laundering
Description & Specifications
Specifications
Palm, wrist
Knit
Nitrile,Polyurethane
Small
1 in.
Level 2
sheet Metal handling, Metal fabrication work
Dyneema™ Fiber
White
2.54cm