Honeywell North™ NorthFlex Light Task Plus II™ Gloves

PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life

Manufacturer: Honeywell NFD15/8M

 View more versions of this product

Catalog No. 19-150-1003

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) sheet Metal handling, Metal fabrication work
    Cut Resistance Rating Level 2
    Length (English) 1 in.
    Length (Metric) 2.54cm
    Material Nitrile, Polyurethane
    Liner Material Dyneema™ Fiber
    Size Medium
    Coated Area Palm, wrist
    Color White

    North NorthFlex Light Task II Polyurethane Coated Dyneema Gloves

    • PU (polyurethane) palm coating is light for comfort and maximum tactility, but provides high abrasion and puncture resistance
    • Glove conforms to the shape of the hand and provides minimal resistance to hand movement, thus greatly reducing fatigue
    • The back of the glove is un-coated, for ventilation and wearer comfort
    • Contains no reaction causing, natural rubber proteins
    • Cuff over-stitching is color-coded by size to aid sorting after laundering

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit