North™ NorthFlex Light Task™ Gloves

PU (polyurethane) palm coating is light for comfort and maximum tactility and provides longer life

Manufacturer: Honeywell Safety Products NF15/10XL

 View more versions of this product

Catalog No. 19-150-0958

  Check Availability
Add to Cart

    Specifications

    Cut Resistance Rating Level 1
    Cleaning Procedure Launderable
    Exterior Finish Sandy
    Glove Cuff Style Knit
    Glove Cuff Color Navy blue
    For Use With (Application) automotive assembly, electronics, general maintenance work, light assembly, manufacturing, small part handling
    Material Polyurethane
    Liner Material Nylon
    Size X-Large
    Coated Area Palm
    Color White

    North NorthFlex Light Task Polyurethane Coated Nylon Gloves

    • Not cut-resistant
    • Polyurethane palm coating provides excellent grip and longer life
    • Uncoated back design allows gloves to breathe for more comfort
    • 15 gauge nylon liner has minimal shrinkage when laundered
    • Cuff over-stitching is color-coded by size to aid sorting after laundering

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit