Learn More
Honeywell™ North™ NorthFlex Oil-Grip™ Gloves
New Oil-Grip gloves are specially designed for heavy-duty/oil applications
Manufacturer:  Honeywell™ NF35/9L
 View more versions of this product
Catalog No. 19-045-565
Add to cart
Description
Description
The unique, nitrile coating is textured with thousands of microscopic pockets that disperse fluids away from the surface of the glove to allow a high level of surface contact between glove and oil saturated object.
  • The MicroFinish™ surface is textured with thousands of microscopic pockets that disperse fluids and allow a high level of surface contact between glove and object
  • MicroFinish goes beyond foamed finishes to provide oily grip that is unsurpassed
  • Two stage coating is impermeable to liquids, so that hands remain dry
  • Lightweight 13 gauge seamless nylon liner and a coating that is soft and flexible, make for an extremely comfortable, form-fitting glove
  • Launderable for added value and longer life
  • Cuff over-stitching is color-coded by size to aid sorting after laundering
Specifications
Specifications
Safety
Knit
Nylon
Dark Green
Textured
Nitrile
Large