North™ NorthFlex Red™ Foamed PVC Palm Coated Gloves

Light and comfortable general purpose gloves that offer high abrasion resistance and exceptional grip

Manufacturer: Honeywell Safety Products NF11/9L

 View more versions of this product

Catalog No. 19-166-667

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    For Use With (Application) agriculture, construction, manufacturing, municipal services, Steel and Metals, utilities
    Material PVC
    Liner Material Nylon
    Size Large
    Coated Area Full palm, finger
    Color Red
    Cleaning Procedure Launderable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit