Learn More
Honeywell™ North™ Norton 180™ T1800 Series Safety Glasses
Molded polycarbonate wraparound safety glasses
Manufacturer:  Honeywell™ 1080H5
 View more versions of this product
Catalog No. 19-068388
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Scratch-resistant lens absorbs 99.9% of UV radiation up to 380nm
  • One-piece, impact-resistant lens offers a large field of vision
  • Frosted browguard reduces glare from overhead lighting
  • Vented sideshields
Compliance
Meet ANSI™ Z87.1-2010 and CSA Z94.3 standards.
Specifications
Specifications
Safety
Norton 180 T1800 Series Safety Glasses
Hardcoated
Eyeglass Pouch