Sperianl™ One-Fit™ N95 Particulate Respirators
Universal sized disposable respirators fit most face shapes and sizes
Manufacturer: Honeywell Safety Products NBW95
 View more versions of this product
Catalog No. 19-999-403
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications NIOSH N95, TC-84A-4373, FDA, CDC
    Quantity 20
    Disposable Yes
    Filter Type N95 Filter
    For Use With (Application) Dusty operation, grinding, polishing, sanding, sweeping, healthcare/medical
    Latex Free Yes
    Material Electrostatic Nonwoven Polypropylene and Polyester
    Size Universal
    Product Type Particulate Respirator
    Construction Lightweight
  • Description & Specifications
Related Products
Product Content Feedback