Learn More
Honeywell Safety Products™ North™ Op-Tema™ Safety Glasses
Manufacturer:  Honeywell Safety Products™ 11150401
 View more versions of this product
Catalog No. 19-039-238
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Lightweight enough for comfortable all-day wear, with adjustable lens angle, temple length, and nose bridge for a secure fit. These general-purpose safety spectacles have a built-in molded browbar for clear, unobstructed vision through the full-view lens. High-impact, abrasion-resistant polycarbonate lens gives 99.9% UV protection. Includes breakaway Flexicord™ retainer for security, convenience.
Compliance
Meet or exceed ANSI Z87.1-1989 and CSA Z94.3-1982 standards.
Specifications
Specifications
Hardcoated
Impact-resistant, Scratch-resistant
Black
TSR Gray
Meet or exceed ANSI Z87.1-1989 and CSA Z94.3-1982 standards.