Learn More
Honeywell Safety Products™ North™ Op-Tema™ Safety Glasses
Specifications
Certifications/Compliance Meet or exceed ANSI Z87.1-1989 and CSA Z94.3-1982 standards.
Lens Coating Hardcoated
Lens Type Impact-resistant, Scratch-resistant
View More Specs
Products 1
Catalog Number Mfr. No. Lens Tint Temple Color Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Temple Color Price Quantity & Availability  
19-039-238
View Documents
Honeywell Safety Products™
11150401
TSR Gray This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Lightweight enough for comfortable all-day wear, with adjustable lens angle, temple length, and nose bridge for a secure fit. These general-purpose safety spectacles have a built-in molded browbar for clear, unobstructed vision through the full-view lens. High-impact, abrasion-resistant polycarbonate lens gives 99.9% UV protection. Includes breakaway Flexicord™ retainer for security, convenience.
Compliance
Meet or exceed ANSI Z87.1-1989 and CSA Z94.3-1982 standards.
Specifications
Specifications
Meet or exceed ANSI Z87.1-1989 and CSA Z94.3-1982 standards.
Hardcoated
Impact-resistant, Scratch-resistant