North Op-Tema™ Safety Glasses
Manufacturer: Honeywell Safety Products 11150400
 View more versions of this product
Catalog No. 19-039-237
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Lens Coating Hardcoat
    Temple Color Black
    Certifications/Compliance Meet or exceed ANSI Z87.1-1989 and CSA Z94.3-1982 standards.
    Lens Material High-impact, Abrasion-resistant Polycarbonate
    Lens Tint Clear
  • Description & Specifications
Related Products
Product Content Feedback