Learn More
Honeywell™ North™ Panther Hip-Pac Respirator
This product provides air from a remote air source for entry into and escape from IDLH atmospheres. The escape cylinder is available in 5-, 10- and 15-minute durations.
Manufacturer:  Honeywell™ P968457
 View more versions of this product
Catalog No. 18-242
Add to cart
Description
Description
The Pressure Demand SAR with Escape Cylinder (Hip-Pac) offers the same reliable facepieces and second stage regulators as the Cougar and 1997-style Panther SCBAs. It allows the user to comfortably wear the respirator to enter an IDLH environment while breathing from a remote air source. Should the air supply be interrupted, the user can switch to the cylinder for escape.
Specifications
Specifications
NIOSH
Chemical, construction, food services, manufacturing, military, mining, oil and gas, pharmaceutical
Hip-Pac Respirator
5 min.
3000psi aluminum cylinder, silicone twenty plus facepiece, regulator, medium, black, medium nose cup, silicone headstrap