Learn More
Honeywell™ North™ Personal Protection Towelettes
Economical towelette delivery systems allow outdoor workers to easily apply protection
Manufacturer:  Honeywell™ 122000X
 View more versions of this product
Catalog No. 19-121-795
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Towelettes come in convenient pocket-sized, durable pouch.
SunX™ Single Dose Packets
  • Contain oil-free, Sun Protection Factor 30 (SPF 30) sunscreen
  • Provide UVA/UVB protection
  • Passed ASTM™ D 412 and D 120 tests regarding effect on lineman's gloves
  • Formula is free of PABA (para-aminobenzoic acid)
  • Perspiration resistant, so skin can cool during strenuous physical activity
  • Easy-to-carry foil pouch comes in dispenser or bulk packs
Bug X™ Towelettes
  • Effective insect repellent DEET (n,n-diethyl-m-toluamide) in towelettes for easy, on-the-spot application
  • Repel mosquitoes, ticks, fleas, mites, black flies, sand flies, and deer flies
  • EPA registered in all 50 states
  • Each 8 x 5 in. (20.3 x 12.7cm) towelette comes in pocket-sized pouch
  • Available in handy dispenser pack
Ivy X™ Pre-Contact Towelettes
  • Almost unnoticeable skin protectant dries quickly, and washes off easily with soap and water
  • Not greasy or sticky
  • Come in dispenser or bulk packs
Ivy X Cleanser Towelettes
  • Unscented towelettes contain moisturizers to be gentle to the skin while scrubbing away poisonous oils
  • Come in dispenser pack
Specifications
Specifications
50 Pack
SunX™ Sunscreen Towelette
Agriculture, construction, fire protection, forestry, government, law enforcement, medical, military, mining, municipal services, Oil and gas, Steel and Metals, transportation, utilities, welding, wind energy