Learn More
Honeywell™ North™ Polar™ Gloves
Thinsulate™ liner provides protection from cold
Manufacturer:  Honeywell™ 70/8210NKY
 View more versions of this product
Catalog No. 19-050-252
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Waterproof vinyl liner prevents hands from getting wet
  • Extended knitwrist provides additional weather protection
  • Gunn cut with wing thumb for comfort
  • Colors to match your favorite sports team
  • Recommended for temperatures down to -36°F (-38°C)
Specifications
Specifications
Safety
cold conditions work including construction, forestry, snow clearing, de-icing, highway work, baggage handling, maintenance work and cold storage
Thinsulate™
-36°F
Knit
Cowhide Leather
Large
Black, Yellow