Learn More
Honeywell™ North™ Polar™ Gloves
Thinsulate™ liner provides protection from cold
Manufacturer:  Honeywell™ 70/8510NKR
 View more versions of this product
Catalog No. 19-050-257
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Waterproof vinyl liner prevents hands from getting wet
  • Extended knitwrist provides additional weather protection
  • Gunn cut with wing thumb for comfort
  • Colors to match your favorite sports team
  • Recommended for temperatures down to -36°F (-38°C)
Specifications
Specifications
Safety
cold storage, packing plants and other low-temperature applications
-38°F
Safety
Cowhide Leather
Gray