Learn More
Honeywell™ North™ Polysafe and Polysafe Plus Wraparound Safety Glasses
Click to view available options
Lens Tint:
Gray
No. per Pack:
72/Cs.
Description
- Sideshields comply with the OSHA Bloodborne Pathogens Standard, so spectacles are ideal for clinical use
- Fit over many types of prescription eyewear
- Polysafe Plus lenses are silicone-coated for scratch resistance and reduced glare
- Polysafe Polycolors spectacles have the same clear lenses as Polysafe spectacles, but with colored temples
- Approximate weight: 39g
Specifications
Lens Tint | Gray |
No. per Pack | 72/Cs. |