North Powdered Latex Finger Cots

Tapered design for more comfortable fit

Manufacturer: Honeywell Safety Products 334LWR/L

 View more versions of this product

Catalog No. 19-063520

  Check Availability
Add to Cart

Alert

Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


    Specifications

    Color White
    Cuff Type Rolled
    Material Natural Rubber
    Thickness (English) 3mil
    Certifications/Compliance ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN374, EN388, EN420, OSHA 29 CFR 1915.157
    Construction Dipped
    Design Tapered
    For Use With (Application) For use in food processing
    For Use With (Equipment) Used in handling delicate components and subminiature parts
    Length (English) 2.75 in.
    Length (Metric) 7cm
    Size Large
    Product Type Finger Cot

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit