North Premier 6100 Half-Mask
Manufacturer: Honeywell Safety Products 313000
 View more versions of this product
Catalog No. 17-632-3
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications NIOSH 42CFR84
    Cartridge Type T Series Cartridge
    For Use With (Application) Chemical, construction, food services, forestry, government, manufacturing, military, mining, municipal services, oil and gas, pharmaceutical, steel and metals, welding
    Includes Dual-cartridge facepiece, nonslip cradle suspension
    Material Silicone
    Weight (English) <0.75 lb.
    Weight (Metric) <340kg
    Product Type North™ Premier 6100 Half Mask
    Size Large
  • Description & Specifications
Related Products
Product Content Feedback