Learn More
North Prevail Safety Glasses
Absorb UV, IR, and blue light radiation
Manufacturer:  Honeywell Safety Products 11150336
 View more versions of this product
Catalog No. 19-087-394B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
TSR Gray
Anti-fog
Meet all ANSI Z87.1-1989 standards, including side impact protection, without the use of conventional sideshields. TSR gray lens tint meets the Traffic Signal Color Recognition requirements of ANSI Z80.3-1986.
Black