Learn More
Honeywell Safety Products™ North™ Prevail Safety Glasses
Absorb UV, IR, and blue light radiation
Specifications
Frame Color Black
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
18-999-2474
View Documents
Honeywell Safety Products™
11150300
N/A Clear lens tint; Uncoated lens; Black Frame This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-087-394B
View Documents
Honeywell Safety Products™
11150336
N/A Gray lens tint; Fog-Ban antifog lens coating; Black Frame This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Sporty, ultra-lightweight design incorporates integrated side impact protection and superior peripheral optics for clear, comfortable vision. Ideal choice for both indoor and outdoor jobs.
Sporty, ultra-lightweight design incorporates integrated side impact protection and superior peripheral optics for clear, comfortable vision.
  • Exclusive, high-performance lenses absorb damaging wavelengths of UV, IR, and blue light radiation
  • Wraparound styling, universal bridge design, and deep curve lenses ensure comfortable fit on virtually any face
  • Unique I/O™ (Indoor/Outdoor) dual-purpose lens reduces glare from artificial and natural light, while providing 100% UV protection
Order Info
Available with Fog-Ban™ antifog coating for clear vision in hot, humid conditions
Compliance
Meet all ANSI Z87.1-1989 standards, including side impact protection, without the use of conventional sideshields. TSR gray lens tint meets the Traffic Signal Color Recognition requirements of ANSI Z80.3-1986.
Specifications
Specifications
Black