Learn More
Honeywell™ Protecto-Shield™ Replacement Windows
Tough faceshield protection with PresLock or Ratchet suspension adjustable headgear. Fits most commonly used visors.
Manufacturer:  Honeywell™ 11390065
 View more versions of this product
Catalog No. 19-166-360
Add to cart
Description
Description
  • Constructed of tough thermoplastic for superior heat protection
  • Designed with a chemical splash drip edge to prevent fluids from splashing down inside the visor
  • Easy sizing and tension adjustment for individual visor positioning
  • Visors easily attach with no snaps, posts or levers
  • Available with ratchet headband with tension knob that allows easy sizing and adjustment for individual visor positioning
  • Available with pinlock headband that provides adjustments similar to a baseball cap
  • Available in propionate, polycarbonate and acetate for a variety of environments
  • Popular industry sizes and thicknesses
  • Easy attachment without snaps, posts or levers and reinforced edge to resist cracking
  • Universal hole pattern allows the use of Sperian visors with other popular manufacturers' headgear
  • Aluminum bound acetate replacement visors form easily for a contoured, customized fit
Compliance
Meets ANSI Z87.1 and CSA Z94.3 standards.
Specifications
Specifications
Clear
8.5 in.
ANSI Z87.1, CSA Z94.3
15 in.
Clear Polycarbonate