North QR Quick Relief™ WoundSeal™ Blood Stopper
Stops bleeding instantly
Manufacturer: Honeywell Safety Products 28QOC051S
 View more versions of this product
Catalog No. 19-310-059
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For occupational workplaces, sports and other applications that require fast treatment of cuts, lacerations, nose bleeds and other external bleeding wounds
    Includes 2 applicators
    Material Topical powder (Hydrophilic polymer and Potassium salt)
    Product Type Blood Stopper Powder
    Quantity 2 applications/box
  • Description & Specifications
Related Products
Product Content Feedback