Learn More
Honeywell™ North™ Series 7900 Emergency Escape Mouthbit Respirator
The 7900 Series mouthbit respirators offer a quick and economical method to escape non-IDLH emergency situations. Use the respirator as a “visitor escape device” or issue individual respirators to employees as a safety precaution against a potential chemical leak
Supplier: Honeywell™ 7904
Description
- Provides escape from acid gas or ammonia
- Convenient neckstrap or beltclip
- Lightweight and disposable
Use for environments where ammonia is present.
Low breathing resistance increases wearer acceptance and reduces fatigue. Exhalation valve prevents inward contamination leakage; inhalation valve keeps exhaled breath and spittle from contacting chemical absorbent area. Break-away neck strap reduces risk of injury should respirator become caught in machinery. Large stainless-steel belt clip ensures easy attachment to belt or pants.
Specifications
Mouthbit Respirator | |
Exhalation Valve; Inhalation Valve; Break-away Neck Strap; Large Stainless-steel Belt Clip |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.