Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ Sinus Relief

Relieve sinus and cold discomfort

$22.25

Specifications

Format Tablet
For Use With (Application) For relief of sinus pain and congestion
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
17-986-446
View Documents
Honeywell™
2106250
Sinus Decongestant Tablet 250 Pack Each for $22.25
Add to cart
 
Description

Description

  • Sinus Tablets: multi-action tablets for relief of sinus pain and congestion
  • Multi-Symptom Cold Tablets: contain nasal decongestant, cough suppressant and non-aspirin pain reliever
  • Cherry Cough Drops: provide soothing relief of scratchy, sore throats to help prevent coughs

Specifications

Specifications

Tablet
For relief of sinus pain and congestion
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit