North Sinus Relief

Relieve sinus and cold discomfort

Manufacturer: Honeywell Safety Products 2106100

 View more versions of this product

Catalog No. 17-986-445

  Check Availability
Add to Cart

    Specifications

    Active Ingredients Phenylephrine Hydrochloride 5mg
    For Use With (Application) For relief of sinus pain and congestion
    Format Tablet
    Quantity 100 Pack
    Product Type Sinus Decongestant Tablet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit