Learn More
North Static Dissipative Latex Finger Cots
Won't weaken with age, rubbing or washing
Manufacturer:  Honeywell Safety Products 145LBR/XL
 View more versions of this product
Catalog No. 19-067515
  Check Availability
Add to cart
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Specifications Description & Specifications
Specifications
Finger Cot
Black
For use in medicals and chemical industries
7cm
Yes
3mil
ANSI/ISEA 105 - ASTM D3389-05, ANSI/ISEA 105 - ASTM F1790-97, EN374, EN388, EN420, OSHA 29 CFR 1915.157
Rolled
2.75 in.
Natural Rubber
XL