Learn More
Honeywell™ Sperian™ Survivair™ Opti-Fit™ APR Respirator
Compatible with S-Series cartridges and filters
Manufacturer:  Honeywell™ 762000
 View more versions of this product
Catalog No. 19-065-188B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Full-face APR respirator provides optimum fit, comfort and superior optics
  • Head harness: 5-point strap
  • Wide viewing area offers distortion free visibility and optical clarity
  • Silicone skirt provides exceptional durability and comfort
  • All straps are attached to rim of facepiece, rather than the silicone skirt, to reduce pressure points and distortion of the sealing surface when tightened
  • Nose cup prevents fogging with optional kits for customized sizing (comes standard with medium nose cup); optional clear nose cup kits available
  • Replaceable, high-impact resistant polycarbonate lens treated with anti-scratch coating
  • Molded in speaking diaphragm provides improved audio transmission for communication
  • Available in three sizes for wide range of fit
  • Hazards: Airborne Particulates, Biohazard, Chemical, Contamination, Extreme Environment, Gas, Vapors, Smoke
Applications:
Agriculture, Chemical, Construction, Fire Protection, Food Services, Forestry, Government, Law Enforcement, Manufacturing, Medical, Military, Mining, Municipal Services, Nuclear, Oil and Gas, Pharmaceutical, Steel and Metals, Transportation, Utilities, Welding.
Compliance
NIOSH 42 CFR 84.
Warranty and Services
Free of defects in materials and workmanship for five years (for the skirt) and one year (for all other components) from date of purchase.
Specifications
Specifications
Safety
S Series and T Series Cartridge
Recommended industries/use : agriculture, chemical, construction, fire protection, food services, forestry, government, law enforcement, manufacturing, medical, military, mining, municipal services, nuclear, oil and gas, pharmaceutical, steel and metals,
Speaking diaphragm
5 point head strap
Polycarbonate
Air Purifying Respirator
Threaded
Survivair™ s series cartridge
Silicone
Medium
NIOSH 42CFR84