North T-Series (6000 and 8000) P100 Combination Filter/Cartridges

Manufacturer: Honeywell Safety Products T105410

 View more versions of this product

Catalog No. 19-003-443A

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Quantity 4 Pack
    For Use With (Equipment) Survivair™ t series respirator,Survivair™ t series respirator
    Product Type Filter/Cartridge
    Certifications NIOSH
    Cartridge Type Filter Cartridge
    Color Green, Magenta

    Tychem SL is Tychem coated with Saranex™ 23-P to provide protection from an even wider range of chemicals. Hoods cover head, neck, and face; they feature a wide-angled plastic lens.

    Double Bib (Inner Bib) Style: Outer bib with a long inner bib; tuck inner bib into shirt or coveralls to funnel excess air from the hood to the body
    Double Bib (Inner Bib) Style for Use with Hardhat: Accommodates Bullard hardhats

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit