Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell North Traffic Control Flags
Tear-proof material
Manufacturer: Honeywell FLA18
Description
- Made from PVC nylon
- Dowel sleeve
- Size: 45.7 x 45.7cm (18.0 in. x 18.0 in.)
- Color: Orange
Specifications
Orange | |
Traffic Flag | |
Dowel sleeve | |
PVC or Nylon |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title