Learn More
Honeywell™ North™ V-Maxx Goggles
Direct- and indirect-vented models with Fog-Ban™ antifog, antiscratch coating for durability
Supplier:  Honeywell™ 11250830
 View more versions of this product
Catalog No. 19-065-518
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Comfortable protection against impact, dust, mist and chemical splash
  • Wraparound design provides maximum coverage and 180° field of vision
  • Soft rubber construction ensures secure, comfortable fit
  • Large enough to be worn over prescription glasses
  • Red headband for impact-resistant vented models; black headband for chemical-splash indirect vented models
Red headband for impact-resistant vented models; black headband for chemical-splash indirect vented models
Order Info
Peel-off goggle covers are available; contact your Fisher Scientific Customer Service Representative.
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Specifications
Specifications
ANSI Z87.1-2010, CSA Z94.3
IR/PC 3.0
Safety
Indirect vent
Elastic
Scratch-resistant, Anti-fog
Goggles
Black
Fog-Ban Anti-fog