Learn More
Honeywell™ North™ V-Maxx Goggles
Catalog Number 19065519
Click to view available options
Product Type:
Goggles
Lens Tint:
Clear
IR/PC 5.0
This item is not returnable. View return policy
This item is not returnable. View return policy
Direct- and indirect-vented models with Fog-Ban™ antifog, antiscratch coating for durability
  • Comfortable protection against impact, dust, mist and chemical splash
  • Wraparound design provides maximum coverage and 180° field of vision
  • Soft rubber construction ensures secure, comfortable fit
  • Large enough to be worn over prescription glasses
  • Red headband for impact-resistant vented models; black headband for chemical-splash indirect vented models
Red headband for impact-resistant vented models; black headband for chemical-splash indirect vented models
Order Info
Peel-off goggle covers are available; contact your Fisher Scientific Customer Service Representative.
Compliance
Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Certifications/Compliance ANSI Z87.1-2010, CSA Z94.3
Lens Tint IR/PC 5.0
Product Type Goggles
For Use With (Application) Safety
Frame Color Black
Ventilation Details Indirect vent
Frame Material Elastic