Learn More
Honeywell™ North™ Vapor Safety Glasses
Click to view available options
Lens Tint:
Clear
TSR Gray
Lens Coating:
Anti-fog
Hardcoat
Description
Capture sports styling in a painted, metallic blue frame
• 9–base wrap-around dual lenses and sport temples offer a comfortable secure fit
• Soft nose bridge prevents slipping
• Durable polycarbonate lens feature scratch-resistant or Fog-Ban™ antifog coating
• Adjustable breakaway Flexicord™ included
Compliance
Meet ANSI Z87+ and CSA Z94.3 safety standards
Specifications
Lens Coating | Hardcoat |
Certifications/Compliance | Meet ANSI Z87+ and CSA Z94.3 safety standards |
Lens Tint | TSR Gray |
Includes | Break-Away Flexicord |
Temple Style | Sport |
Lens Material | Polycarbonate |