Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nortropinone hydrochloride, ≥98%

Click to view available options
Quantity:
5g
25g
100g
25602-68-0
C7H12ClNO
161.629
MFCD03613582
MZQWQFWRSDNBPV-UHFFFAOYSA-N
nortropinone hydrochloride,8-azabicyclo 3.2.1 octan-3-one hydrochloride,nortropinone hcl,8-azabicyclo 3.2.1 octan-3-one hcl,8-azabicyclo 3.2.1 octan-3-one, hydrochloride,3-nortropanone hydrochloride,8-aza-3-oxobicyclo 3.2.1 octane hydrochloride,nortropinonehydrochloride,c7h11no.clh,8-azabicyclo 3,2,1 octan-3-one hydrochloride
13091218
8-azabicyclo[3.2.1]octan-3-one;hydrochloride
C1CC2CC(=O)CC1N2.Cl
25602-68-0
C7H12ClNO
161.629
MFCD03613582
MZQWQFWRSDNBPV-UHFFFAOYSA-N
nortropinone hydrochloride,8-azabicyclo 3.2.1 octan-3-one hydrochloride,nortropinone hcl,8-azabicyclo 3.2.1 octan-3-one hcl,8-azabicyclo 3.2.1 octan-3-one, hydrochloride,3-nortropanone hydrochloride,8-aza-3-oxobicyclo 3.2.1 octane hydrochloride,nortropinonehydrochloride,c7h11no.clh,8-azabicyclo 3,2,1 octan-3-one hydrochloride
13091218
8-azabicyclo[3.2.1]octan-3-one;hydrochloride
C1CC2CC(=O)CC1N2.Cl
Melting Point 194°C to 196°C
Color Undesignated
Sensitivity Hygroscopic
Quantity 5g
Beilstein 3693524
Formula Weight 161.63
Physical Form Solid
Percent Purity ≥98%
Chemical Name or Material Nortropinone hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C