Learn More
BioVision IncSupplier Diversity Partner Nitric Oxide Synthase (NOS) Activity Assay Kit (Colorimetric): Nitric Oxide Synthase (NOS) Activity Assay Kit (Colorimetric): Ultrasensitive, Simple, HTP adaptable assay kit for NOS activity measurement. 100 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K205100
Nitric Oxide Synthase (NOS) Activity Assay Kit (Colorimetric): Nitric oxide synthases (EC 1.14.13.39) (NOS) are a family of enzymes that catalyze the production of nitric oxide (NO) from L-arginine. Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. There are three isoforms of NOS: endothelial (eNOS), neuronal (nNOS), and inducible (iNOS). BioVision's Nitric Oxide Synthase Activity Assay Kit provides an accurate and convenient method to assay NOS activity in a variety of samples. In this assay, nitric oxide generated by NOS undergoes a series of reactions and reacts with Griess Reagent 1 and 2 to generate a colored product with a strong absorbance at 540 nm. The assay is simple, sensitive and high-throughput adaptable and can detect as low as 5 µU of NOS activity. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9617