Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOSTRIN Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548643

Catalog No. PA548643

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Multivalent adapter protein which may decrease NOS3 activity by inducing its translocation away from the plasma membrane.
Specifications

Specifications

NOSTRIN
Polyclonal
PBS with 0.09% sodium azide
Q8IVI9, Q6WKZ7
Nostrin, BM247 homolog, Nitric oxide synthase traffic inducer, Nitric oxide synthase trafficker, eNOS-trafficking inducer, NOSTRIN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
115677, 329416
Western Blot
Unconjugated
NOSTRIN
Liquid
NOSTRIN
KLH conjugated synthetic peptide between 296-325 amino acids from the Central region of human NOSTRIN.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit