Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch-2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093549AF700
Description
Notch-2 Polyclonal antibody specifically detects Notch-2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
Notch-2 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
Alexa Fluor 700 | |
AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2 | |
Synthetic peptide representing the C terminal region of the human NOTCH homolog 2 (NOTCH2) protein. | |
0.1 mL | |
Apoptosis, Cancer, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Signal Transduction, Stem Cell Signaling Pathway | |
4853 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction