Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00
Specifications
| Antigen | Notch-2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017905
![]() |
Novus Biologicals
NB10093549SS |
0.025 mg |
Each for $206.00
|
|
|||||
NB10093549
![]() |
Novus Biologicals
NB10093549 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Notch-2 Polyclonal specifically detects Notch-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| Notch-2 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 4853 | |
| Synthetic peptide representing the C terminal region of the human NOTCH homolog 2 (NOTCH2) protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| RUO | |
| Human | |
| AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2 | |
| NOTCH2 | |
| IgG | |
| Affinity Purified | |
| Notch 2 homolog |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title