Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch-4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310954100UL
Description
Notch-4 Polyclonal specifically detects Notch-4 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Notch-4 | |
Polyclonal | |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
neurogenic locus notch homolog protein 4, notch 4, Notch homolog 4, NOTCH3 | |
The immunogen is a synthetic peptide directed towards the middle region of human Notch-4. Peptide sequence LLLGLGAARELRDQAGLAPADVAHQRNHWDLLTLLEGAGPPEARHKATPG | |
100 μg | |
Angiogenesis | |
4855 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction