Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTCH3 Armenian Hamster anti-Mouse, APC, Clone: HMN3-133, eBioscience™ Non-distribution product as customer accommodation.

Armenian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen 17576380

 View more versions of this product

Catalog No. 50-112-3137


Add to cart

Description

Description

Description: This HMN3-133 monoclonal antibody reacts with mouse Notch3, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription. Notch 3 expression has been demonstrated on regulatory T cells and some thymocyte subsets, including CD4-CD8- and CD8SP cells. In addition, this Notch receptor is prevalent in vascular smooth muscle and the central nervous system. In addition to its role in stem cell hematopoiesis, Notch 3 plays a pivotal role in mammalian T cell lineage commitment and thymocyte development. Specifically, Notch 3 has been implicated in the CD4-CD8- double negative (DN) to CD4+CD8+ double positive transition with greatest expression in DN3 (CD4-CD8-CD25+CD44-) cells. Notch 3 may also be involved in regulating the pre-TCR checkpoint via crosstalk with the NFkappaB and E2A signaling pathways.

Finally, overexpression of Notch 3 has been implicated in leukemia. Applications Reported: This HMN3-133 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HMN3-133 antibody has been tested by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered. NOTCH3 is the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal domit arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).
Specifications

Specifications

Notch3
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q61982
CADASIL, CASIL
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
HMN3-133
APC
Notch3
Liquid
Notch3
IgG
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
18131
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.