Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTCH4 Mouse anti-Human, Clone: L5C5, Abnova™

Mouse monoclonal antibody raised against human NOTCH4.

Manufacturer:  Abnova Corporation MAB12688

Catalog No. 89-934-705


Add to cart

Description

Description

This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. The Notch signaling network is an evolutionarily conserved intercellular signaling pathway which regulates interactions between physically adjacent cells. In Drosophilia, notch interaction with its cell-bound ligands (delta, serrate) establishes an intercellular signaling pathway that plays a key role in development. Homologues of the notch-ligands have also been identified in human, but precise interactions between these ligands and the human notch homologues remain to be determined. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. This gene may be associated with susceptibility to schizophrenia in a small portion of cases. An alternative splice variant has been described but its biological nature has not been determined. [provided by RefSeq]

Specifications

Specifications

NOTCH4
L5C5
Mouse monoclonal antibody raised against human NOTCH4.
In 10mM sodium HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 0.1mg/mL BSA, 0.02% sodium azide).
FLJ16302/INT3/MGC74442/NOTCH3
Mouse
IgG1
RUO
Primary
4855
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Immunoprecipitation (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
NOTCH4
Recombinant protein corresponding to human NOTCH4.
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit