Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOV Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554055
Description
Immunogen sequence: SCCLVCARQR GESCSDLEPC DESSGLYCDR SADPSNQTGI CTAVEGDNCV FDGVIYRSGE KFQPSCKFQC TCRDGQIGC Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
NOV is a member of the CCN family of secreted, cysteine-rich regulatory proteins. The full-length NOV protein contains four structural domains that confer distinct, and sometimes opposing, biological activities. Elevated expression of NOV is associated with certain tumors, including Wilm's tumor and most nephroblastomas. However, in other tumor types and certain cancer cell lines, increased tumorigenicity and proliferation is correlated with decreased NOV expression. Additionally, NOV induces cell adhesion and cell migration by signaling through specific cell-surface integrins, and by binding to heparin sulfate proteoglycans and to fibulin 1C. NOV has also been reported to exert proangiogenic activities.
Specifications
NOV | |
Polyclonal | |
Unconjugated | |
CCN3 | |
C130088N23Rik; CCN family member 3; CCN3; Cellular communication network factor 3; IBP-9; IGF-binding protein 9; IGFBP9; IGFBP-9; Insulin-like growth factor-binding protein 9; Nephro blastoma-overexpressed gene protein homolog; nephroblastoma overexpressed; nephroblastoma overexpressed gene; nephroblastoma overexpressed gene protein homolog; nephroblastoma-overexpressed gene protein homolog; NOV; NovH; protein NOV homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4856 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P48745 | |
CCN3 | |
Recombinant protein corresponding to Human NOV. Recombinant protein control fragment (Product #RP-91870). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction