Learn More
Invitrogen™ NOV Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579750
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell. IHC: human intestinal cancer tissue, human lung cancer tissue, human mammary cancer tissue, rat kidney tissue. ICC/IF: A431 cell. Flow: U87 cell.
NOV is a member of the CCN family of secreted, cysteine-rich regulatory proteins. The full-length NOV protein contains four structural domains that confer distinct, and sometimes opposing, biological activities. Elevated expression of NOV is associated with certain tumors, including Wilm's tumor and most nephroblastomas. However, in other tumor types and certain cancer cell lines, increased tumorigenicity and proliferation is correlated with decreased NOV expression. Additionally, NOV induces cell adhesion and cell migration by signaling through specific cell-surface integrins, and by binding to heparin sulfate proteoglycans and to fibulin 1C. NOV has also been reported to exert proangiogenic activities.
Specifications
NOV | |
Polyclonal | |
Unconjugated | |
CCN3 | |
C130088N23Rik; CCN family member 3; CCN3; Cellular communication network factor 3; IBP-9; IGF-binding protein 9; IGFBP9; IGFBP-9; Insulin-like growth factor-binding protein 9; Nephro blastoma-overexpressed gene protein homolog; nephroblastoma overexpressed; nephroblastoma overexpressed gene; nephroblastoma overexpressed gene protein homolog; nephroblastoma-overexpressed gene protein homolog; NOV; NovH; protein NOV homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4856, 81526 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
P48745, Q9QZQ5 | |
CCN3 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human NOV/CCN3 (334-357aa HTNCPKNNEAFLQELELKTTRGKM). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.