Learn More
High-Tech Conversions NOVA-TECH™ NT1 Nonwoven Cleanroom Wipes
High-Tech Conversions NOVA-TECH™ NT1 Nonwoven Cleanroom Wipes for ISO 5-6 (Class 100-1000) Controlled Environments
Manufacturer:  High-Tech Conversions NT177/DEL
 View more versions of this product
Catalog No. 50-105-9546
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Poly-Cellulose fabric blend provides a tough, absorbent and abrasion resistant product
  • Compatible with virtually any solvent or solution for cleaning and wiping
  • Meets USP 797 and USP 800 Standards
  • Double bag packaging
Specifications
Specifications
18 cm
68 gsm
7 x 7 in.
18 cm
Cleanroom Wiper
Non-sterile
No
White
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Poly-Cellulose Blend
Yes