Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions NOVA-TECH™ NT1 Nonwoven Cleanroom Wipes

High-Tech Conversions NOVA-TECH™ NT1 Nonwoven Cleanroom Wipes for ISO 5-6 (Class 100-1000) Controlled Environments

Specifications

Autoclavable No
Basis Weight 68 gsm
Color White
Disposable Yes
For Use With (Application) Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
50-105-9545
View Documents
High-Tech Conversions
NT166/DEL
6 x 6 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
50-105-9546
View Documents
High-Tech Conversions
NT177/DEL
7 x 7 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Poly-Cellulose fabric blend provides a tough, absorbent and abrasion resistant product
  • Compatible with virtually any solvent or solution for cleaning and wiping
  • Meets USP 797 and USP 800 Standards
  • Double bag packaging
Specifications

Specifications

No
White
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Cleanroom Wiper
68 gsm
Yes
Poly-Cellulose Blend
Non-sterile
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit