Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ NovaBiochem™ Deblock Reagent, 10% DCA in Toluene
Used for synthesis of DNA
$336.00 - $2435.00
Specifications
Color | Clear |
---|---|
Concentration | 10% (v/v) |
Product Type | DNA labeling reagents |
Synonym | DCA in Toluene |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MBI08414000
|
MilliporeSigma™
BI08414000 |
4 L |
Each for $336.00
|
|
|||||
MBI0841NP20
|
MilliporeSigma™
BI0841NP20 |
20 L |
Each for $2,435.00
|
|
|||||
Description
- Used in labeling, biosynthesis of DNA
Specifications
Clear | |
DNA labeling reagents | |
Liquid |
10% (v/v) | |
DCA in Toluene |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title