Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Bacteriophage CE6

Contains T7 RNA polymerase gene.

$194.00 - $750.00

Specifications

Product Type Recombinant Lambda Phage
Storage Requirements ≤−70°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
69-390-3
View Documents
MilliporeSigma™
693903
0.2 mL
Each for $194.00
Add to Cart
 
69-390-4
View Documents
MilliporeSigma™
693904
10 mL
Each for $750.00
Add to Cart
 
Description

Description

EMD Millipore Novagen™ Bacteriophage CE6 is recombinant lambda phage.

  • Recombinant lambda phage
  • Provides source of T7 RNA polymerase to susceptible host cells carrying T7 expression plasmids
  • Has λcI857 immunity Sam7 mutation, which requires supF hosts for lytic growth
  • Provided as high titer lysate includes host strains LE392 (rK12- mK12+) ED8739 (rK12- mK12-) as glycerol stocks
  • 10mL size can induce up to 100mL culture
Specifications

Specifications

Recombinant Lambda Phage
≤−70°C
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit