Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Insect GeneJuice™ Transfection Reagent

Optimized for maximal transfection efficiency of Sf9 insect cells for baculovirus protein expression.

$1500.00

Specifications

Format Liquid
View More Specs

Products 3
Catalog Number Mfr. No. Storage Requirements Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Storage Requirements Quantity Price Quantity & Availability  
71-259-5 View Documents MilliporeSigma™
712595
2° to −8°C 10mL Each for $1,500.00
Add to cart
 
71-259-4 View Documents MilliporeSigma™
712594
2° to −8°C 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
71-259-3 View Documents MilliporeSigma™
712593
2° to 8°C 0.3mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

EMD Millipore Novagen™ Insect GeneJuice™ Transfection Reagent used for both transient and stable transfections.

    Low toxicity to cells

Applications
:
  • Used for transfection of transfer plasmids with linearized virus DNA
  • For reproduction of recombinant baculovirus
  • For large-scale protein expression in suspension-culture transfections of Sf9 insect cells
Specifications

Specifications

Liquid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit